4,4,4-Trifluorobutyraldehyde hydrate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 4,4,4-Trifluorobutanal hydrate
CAS Number:1448773-75-8
:
Formula:C₄H₇F₃O₂
Molecular Weight:144.09 g/mol
Storage Temperature:Refrigerator
UN:1993
ADR:3 III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together