Order Entry
Belgium
ContactUsLinkComponent
Lithium hexafluorophosphate 95%
Corrosive
Toxic
Lithium hexafluorophosphate 95%
Catalog # APOSPC5000-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Toxic
Lithium hexafluorophosphate 95%
Catalog # APOSPC5000-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Nalgene bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Melting Pt:200 °C (dec.)
Density:1.5 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2928
ADR:6.1+8,II
 :151.91 g/mol
EINECS:244-334-7
 :GHS05
 :GHS06
CAS Number:21324-40-3
Formula:F₆LiP
MDL Number:MFCD00011096