Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Fluorophenol 99%

4-Fluorophenol 99%

Corrosive
Toxic
Supplier:   Apollo Scientific
4-Fluorophenol 99%
Supplier:  Apollo Scientific
Certificates
4-Fluorophenol 99%

3 Options Available Below

Product Details & Documents
Synonyms:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185 °C
  • Melting Pt:44…47 °C
  • Density:1.22 g/cm³ (25 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:206-736-0
  • UN:2928
  • ADR:6.1+8,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 23,20 €
    23,20 €
    1 * 100 g
       23,20 €

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 68,10 €
    68,10 €
    1 * 500 g
       68,10 €

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 132,00 €
    132,00 €
    1 * 1 kg
       132,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.