4-Hydroxy-4-(trifluoromethyl)cyclohexanecarboxylic acid 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1163729-47-2
:
Formula:C₈H₁₁F₃O₃
MDL Number:MFCD13659409
:212.1700 g/mol
Storage Temperature:Ambient
Frequently Bought Together