3-Chloro-4-fluorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:91170-93-3
:
Formula:C₆H₃Cl₂FO₂S
MDL Number:MFCD00041444
Boiling Pt:104…105 °C/1mmHg
Density:1.61 g/cm³ (25 °C)
Flash Pt:110 °C
UN:3265
ADR:8,II
:229.06 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together