Boc-3-(trifluoromethyl)-D-phenylalanine
Catalog # APOSPC0043-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:333.31 g/mol
Melting Pt:135…138 °C
Storage Temperature:Refrigerator
CAS Number:82317-82-6
Formula:C₁₅H₁₈F₃NO₄
MDL Number:MFCD00672525