2,4-Heptadien-6-one 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 5-Acetyl-2,4-pentadiene
CAS Number:3916-64-1
:
Formula:C₇H₁₀O
MDL Number:MFCD00059238
Boiling Pt:88 °C
Density:0.9 g/cm³ (25 °C)
Molecular Weight:110.16 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together