Order Entry
Belgium
ContactUsLinkComponent
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
Catalog # APOSOR911731-100G
Supplier:  Apollo Scientific
CAS Number:  
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
Catalog # APOSOR911731-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :166.1739 g/mol
Storage Temperature:Ambient
CAS Number:7326-19-4
Formula:C₉H₁₀O₃
MDL Number:MFCD00078062