trans-4-Hydroxy-L(-)-proline
Bottle, 100 g
Catalog # APOSOR6733-100G
Supplier: Apollo Scientific
trans-4-Hydroxy-L(-)-proline
Catalog # APOSOR6733-100G
Supplier: Apollo Scientific
CAS Number: 51-35-4
Jump to:
Product Details & Documents
Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
- CAS Number:51-35-4
- Formula:C₅H₉NO₃
- MDL Number:MFCD00064320
- Molecular Weight:131.13 g/mol
- Melting Pt:270…273 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-091-9
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:100 g



