

Cyclopentyl methyl ether (CPME) 99% stabilised
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5614-37-9
:
Formula:C₆H₁₂O
MDL Number:MFCD08276401
:100.16 g/mol
Boiling Pt:106 °C
Melting Pt:–134 °C
Density:0.86 g/cm³ (25 °C)
Flash Pt:–1 °C
Storage Temperature:Ambient
UN:3271
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together