Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanillin 97+%

Vanillin 97+%

Irritant
New Product
Supplier:   Apollo Scientific
Vanillin 97+%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:170 °C/15mmHg
  • Melting Pt:81…83 °C
  • Density:1.056 g/cm³ (25 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2

Documents

Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 24,60 €
    24,60 €
    1 * 100 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 34,80 €
    34,80 €
    1 * 500 g
       34,80 €

    Specifications:
    New Product
    New Product