2-Methylpyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-Boc-2-methyl-2-pyrrolidinecarboxylic acid, 1-Boc-2-methyl-DL-proline
CAS Number:203869-80-1
:
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD03095494
Molecular Weight:229.28 g/mol
Melting Pt:104…106 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together