Methyl-3-nitrosalicylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22621-41-6
:
Formula:C₈H₇NO₅
MDL Number:MFCD00272263
:197.15 g/mol
Melting Pt:127…132 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together