Order Entry
Belgium
ContactUsLinkComponent
3-(4-Bromophenyl)-5-chloro[1,2,4]thiadiazole
3-(4-Bromophenyl)-5-chloro[1,2,4]thiadiazole
Catalog # APOSOR346103-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Bromophenyl)-5-chloro[1,2,4]thiadiazole
Catalog # APOSOR346103-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :275.56 g/mol
Storage Temperature:Ambient
CAS Number:191919-26-3
Formula:C₈H₄BrClN₂S
MDL Number:MFCD11052445