2-Bromophenyl isothiocyanate 98%
Catalog # APOSOR3080-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:214.09 g/mol
Boiling Pt:257 °C
Melting Pt:20 °C
Density:1.591 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:13037-60-0
Formula:C₇H₄BrNS
MDL Number:MFCD00004799