
Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Catalog # APOSOR307093-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.27 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:288159-40-0
Formula:C₁₀H₁₉NO₅
MDL Number:MFCD02682576