4-Methoxyphenyl isothiocyanate 97%
Catalog # APOSOR30146-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:165.22 g/mol
Boiling Pt:280…281 °C
Melting Pt:18 °C
Density:1.196 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:218-921-3
UN:1760
ADR:8,II
:GHS05
:GHS07
CAS Number:2284-20-0
Formula:C₈H₇NOS
MDL Number:MFCD00011676