
5-Hydroxy-2-nitrobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:610-37-7
:
Formula:C₇H₅NO₅
MDL Number:MFCD00017566
:183.12 g/mol
Melting Pt:168…172 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together