

3-Hydroxy-4-iodobenzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:58123-77-6
:
Formula:C₇H₅IO₃
MDL Number:MFCD02068386
:264.02 g/mol
UN:2811
ADR:6.1,III
:GHS05
:GHS06
Storage Temperature:Ambient
Frequently Bought Together