Order Entry
Belgium
Orders LinkContactUsLinkComponent
Curcumin 97%

Curcumin 97%

Irritant
Supplier:   Apollo Scientific
Curcumin 97%
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
  • CAS Number:458-37-7
  • Formula:C₂₁H₂₀O₆
  • MDL Number:MFCD00008365
  • Molecular Weight:368.39 g/mol
  • Melting Pt:170…175 °C
  • Storage Temperature:Freezer
  • EINECS:207-280-5

Documents

Product Family Options

Product Information