
2'-Hydroxy-4',6'-dimethoxyacetophenone 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:90-24-4
:
Formula:C₁₀H₁₂O₄
MDL Number:MFCD00017243
:196.2
Melting Pt:80…82 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together