Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Mesitylene

Flammable
Irritant
Environment
Supplier:   Apollo Scientific
Mesitylene
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Boiling Pt:163…166 °C
  • Melting Pt:–45 °C
  • Density:0.864 g/cm³ (25 °C)
  • Flash Pt:53 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 23,20 €
    23,20 €
    1 * 100 mL
       23,20 €

  • Bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 63,70 €
    63,70 €
    1 * 500 mL
       63,70 €

  • Bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 258,00 €
    258,00 €
    1 * 2,5 L
       258,00 €