3-(1H-1,2,4-Triazol-1-yl)-2-butanone
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-(3-Oxobut-2-yl)-1H-1,2,4-triazole
CAS Number:111451-34-4
:
Formula:C₆H₉N₃O
MDL Number:MFCD00142027
:139.16 g/mol
Boiling Pt:93…95 °C/0.38mmHg
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together