Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-2-methylphenylboronic acid
4-Hydroxy-2-methylphenylboronic acid
4-Hydroxy-2-methylphenylboronic acid
4-Hydroxy-2-methylphenylboronic acid
APOSOR10497-1G
:
Apollo Scientific
:
4-Hydroxy-2-methylphenylboronic acid
APOSOR10497-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
151.96 g/mol
:
100…104 °C
:
GHS07
:
Ambient
:
493035-82-8
:
C₇H₉BO₃
:
MFCD03788424