Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB)

3,3',5,5'-Tetramethylbenzidine (TMB)

Bottle
Catalog # APOSOR10053-1G
Irritant
New Product
Supplier:   Apollo Scientific
3,3',5,5'-Tetramethylbenzidine (TMB)
Catalog # APOSOR10053-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB)

Bottle

CAS Number:  54827-17-7
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Melting Pt:168…171 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.