3-Chloro-2-methylphenyl isocyanate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40397-90-8
:
Formula:C₈H₆ClNO
MDL Number:MFCD00037036
:167.59 g/mol
Boiling Pt:94…95 °C/9mmHg
Density:1.247 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Refrigerator
UN:2206
ADR:6.1,III
:GHS08
Frequently Bought Together