Order Entry
Belgium
ContactUsLinkComponent
Palladium(II) acetate trimer
Corrosive
Irritant
Palladium(II) acetate trimer
Catalog # APOSIN2803-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
Palladium(II) acetate trimer
Catalog # APOSIN2803-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :673.52 g/mol
Melting Pt:205 °C
EINECS:258-424-9
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:53189-26-7
Formula:C₁₂H₁₈O₁₂Pd₃