Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Valinomycin

Toxic
Supplier:   Apollo Scientific
Valinomycin
Supplier:  Apollo Scientific
Certificates

6 Options Available Below

Product Details & Documents

Anitbiotic produced by Streptomyces fulvissimus. Valinomycin is highly selective for K+ ions over Na+without influencing the permeability of other ions

  • CAS Number:2001-95-8
  • Formula:C₅₄H₉₀O₁₈N₆
  • MDL Number:MFCD00005114
  • Molecular Weight:1111.34 g/mol
  • Melting Pt:187…190 °C
  • Storage Temperature:Refrigerator
  • EINECS:217-896-6
  • UN:2811
  • ADR:6.1,I
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 65,20 €
    65,20 €
    1 * 25 mg
       65,20 €

  • Bottle
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 118,00 €
    118,00 €
    1 * 50 mg
       118,00 €

  • Bottle
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 209,00 €
    209,00 €
    1 * 100 mg
       209,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 455,00 €
    455,00 €
    1 * 250 mg
       455,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 817,00 €
    817,00 €
    1 * 500 mg
       817,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 470,00 €
    1 470,00 €
    1 * 1 g
       1 470,00 €