Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bis-PEG3-NHS ester

New Product
Supplier:   Apollo Scientific
Bis-PEG3-NHS ester
Supplier:  Apollo Scientific
New Product
Certificates
Bis-PEG3-NHS ester

3 Options Available Below

Product Details & Documents
Synonyms:  Bis-dPEG®3-NHS ester
  • CAS Number:1314378-16-9
  • Formula:C₁₈H₂₄N₂O₁₁
  • Molecular Weight:444.39 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 223,00 €
    223,00 €
    1 * 250 mg
       223,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 507,00 €
    507,00 €
    1 * 1 g
       507,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 2 050,00 €
    2 050,00 €
    1 * 5 g
       2 050,00 €

    Specifications:
    New Product
    New Product