Order Entry
Belgium
Orders LinkContactUsLinkComponent
α-Cyclodextrin

α-Cyclodextrin

Supplier:   Apollo Scientific
α-Cyclodextrin
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 123,00 €
    123,00 €
    1 * 50 g
       123,00 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 174,00 €
    174,00 €
    1 * 100 g
       174,00 €

  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 319,00 €
    319,00 €
    1 * 250 g
       319,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.