Order Entry
Belgium
Orders LinkContactUsLinkComponent
Erythromycin

Erythromycin

Irritant
Supplier:   Apollo Scientific
Erythromycin
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents

Erythromycin is a macrolide antibiotic with a bacteriostatic action against primarily gram-positive bacteria.

  • 960mcg/mg
Synonyms:  (-)-Erythromycin
  • CAS Number:114-07-8
  • Formula:C₃₇H₆₇NO₁₃
  • MDL Number:MFCD00084654
  • Molecular Weight:733.95 g/mol
  • Melting Pt:133 °C
  • Storage Temperature:Ambient
  • EINECS:204-040-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 65,20 €
    65,20 €
    1 * 10 g
       65,20 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 98,40 €
    98,40 €
    1 * 25 g
       98,40 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 274,00 €
    274,00 €
    1 * 100 g
       274,00 €