Order Entry
Belgium
Orders LinkContactUsLinkComponent
n-Decyl-β-D-maltoside

n-Decyl-β-D-maltoside

Irritant
Supplier:   Apollo Scientific
n-Decyl-β-D-maltoside
Supplier:  Apollo Scientific
Certificates
n-Decyl-β-D-maltoside

3 Options Available Below

Product Details & Documents
Synonyms:  DM, N-Decyl-Beta-Maltoside, Decyl 4-O-a-D-glucopyranosyl-b-D-glucopyranoside, n-Decyl-beta-D-maltopyranoside, Decyl b-D-maltopyranoside
  • CAS Number:82494-09-5
  • Formula:C₂₂H₄₂O₁₁
  • MDL Number:MFCD00061624
  • Molecular Weight:482.56 g/mol
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 102,00 €
    102,00 €
    1 * 1 g
       102,00 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 362,00 €
    362,00 €
    1 * 5 g
       362,00 €

  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 615,00 €
    615,00 €
    1 * 10 g
       615,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.