Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Imidazole

Corrosive
Irritant
Health
Supplier:   Apollo Scientific
Imidazole
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents

Biological buffer - useful range 6.2-7.8

  • Assay (by titration) 99%
  • White crystals
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Boiling Pt:235 °C
  • Melting Pt:85…91 °C
  • Density:1.03 g/cm³ (25 °C)
  • Flash Pt:145 °C
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:3263
  • ADR:8,II

 

Product Family Options

Product Information