Order Entry
Belgium
Orders LinkContactUsLinkComponent
γ-Cyclodextrine 98%

γ-Cyclodextrine 98%

Irritant
Supplier:   Apollo Scientific
γ-Cyclodextrine 98%
Supplier:  Apollo Scientific
Certificates
γ-Cyclodextrine 98%

3 Options Available Below

Product Details & Documents
Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 46,40 €
    46,40 €
    1 * 25 g
       46,40 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 121,00 €
    121,00 €
    1 * 100 g
       121,00 €

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 397,00 €
    397,00 €
    1 * 500 g
       397,00 €