Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Galactitol

New Product
Supplier:   Apollo Scientific
Galactitol
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:608-66-2
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064288
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 37,70 €
    37,70 €
    1 * 25 g
       37,70 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 65,20 €
    65,20 €
    1 * 100 g
       65,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 228,00 €
    228,00 €
    1 * 500 g
       228,00 €

    Specifications:
    New Product
    New Product