Order Entry
Belgium
ContactUsLinkComponent
Blasticidin S hydrochloride
Blasticidin S hydrochloride
Catalog # APOSBIB4432-50MG
Supplier:  Apollo Scientific
CAS Number:  
Blasticidin S hydrochloride
Catalog # APOSBIB4432-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
Melting Pt:218…225 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,II
 :458.904 g/mol
 :GHS06
CAS Number:3513-03-9
Formula:C₁₇H₂₇ClN₈O₅
MDL Number:MFCD02091640