Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Aspartic acid

D-(-)-Aspartic acid

Irritant
New Product
Supplier:   Apollo Scientific
D-(-)-Aspartic acid
Supplier:  Apollo Scientific
New Product
Certificates
D-(-)-Aspartic acid

6 Options Available Below

Product Details & Documents

Chiral intermediate for pharmaceuticals and agrochemicals; Materials for peptides.

Synonyms:  D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
  • CAS Number:1783-96-6
  • Formula:C₄H₇NO₄
  • MDL Number:MFCD00063081
  • Molecular Weight:133.1 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Density:1.66 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:217-234-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 21,80 €
    21,80 €
    1 * 100 g
       21,80 €

  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 33,30 €
    33,30 €
    1 * 250 g
       33,30 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 57,90 €
    57,90 €
    1 * 500 g
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 112,00 €
    112,00 €
    1 * 1 kg
       112,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 550,00 €
    550,00 €
    1 * 5 kg
       550,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 1 090,00 €
    1 090,00 €
    1 * 10 kg
       1 090,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.