Order Entry
Belgium
Orders LinkContactUsLinkComponent
Xylan (from beechwood)

Xylan (from beechwood)

Irritant
Supplier:   Apollo Scientific
Xylan (from beechwood)
Supplier:  Apollo Scientific
Certificates
Xylan (from beechwood)

3 Options Available Below

Product Details & Documents
  • CAS Number:9014-63-5
  • MDL Number:MFCD00082148
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:232-760-6

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.