Orders LinkContactUsLinkComponent
SafetyCaps
 :   S.C.A.T.
SafetyCaps
 :  S.C.A.T.
SafetyCaps
SafetyCaps
The SCAT SafetyCaps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. Complete with integrated air valve for pressure equalisation.
  • High quality materials ensure maximum safety for lab and staff
  • 360° rotatable and up to 200 °C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of the connections
  • Improved design for easy connection of capillaries in various colours
  • Free air vaive with change indicator included with every SafetyCap
SCAT SafetyCaps are available in the following versions:
SafetyCaps, basic: Standard SafetyCap with different connections without additional functions.
SafetyCaps, preparative: For use in the preparative HPLC environment with larger tubing connections and air valves with a higher flow rate.
SafetyCaps, with shut-off: Prevent air pockets in the tube when the supply bottle is being changed and guarantees that the analysis continues without a problem.
SafetyCaps, combined: Provides a stable baseline and reliable reproducibility for RI detection for a fast and simple solvent degassing with helium.
SafetyCaps, angled: With lateral connections for capillary and air valve in case of limited space above the bottle.
SafetyCaps, for Merck® bottles: Connect containers with thread sizes S 40 and GL 40 directly to the HPLC system, without decanting or thread adapters.
SafetyCaps, for NS 29/32 ground neck bottles: The locknut allows the stopper to be easily loosened and removed for bottles with NS 29/32 mm standard ground joints.
 : All SafetyCaps are delivered with an air valve and corresponding fittings. PFA fittings, PTFE body and PPS screw cap are conforming to the requirements of flammability classification V-0 (UL-94). Blind plugs are provided to tighten connections which are temporarily inactive or not in use.
Product Family Options
Basic
  • Description
    Thread
    Connector
  • SafetyCap II
    GL32
    2× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  • SafetyCap I, V2.0
    GL45
    1× PFA-fitting 1,6/2,3/3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 98,50 €
    98,50 €
    1 * 1 ST
       98,50 €
  • SafetyCap II
    GL28
    2× PFA-fitting 1,6/2,3/3,2 mm OD, 1× PFA-blind plug
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  • SafetyCap II, V2.0
    GL45
    2× PFA-fitting 1,6/2,3/3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €
  • SafetyCap III, V2.0
    GL45
    3× PFA-fitting 1,6/2,3/3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €
  • SafetyCap I
    GL80
    1× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • SafetyCap IV, V2.0
    GL45
    4× PFA-fitting 1,6/2,3/3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • SafetyCap VI, V2.0
    GL45
    6× PFA-fitting 1,6/2,3/3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • SafetyCap II
    GL80
    2× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €
  • SafetyCap II
    B53
    2× PFA fitting 1.6/2.3/3.2 mm OD
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €
  • SafetyCap III
    GL38
    3× PFA-fitting 1,6/2,3/3,2 mm OD, 2× PFA blind plug
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  • SafetyCap III
    GL80
    3× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • SafetyCap IV
    83B
    4× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 268,00 €
    268,00 €
    1 * 1 ST
       268,00 €
For special bottles
  • Description
    Thread
    Connector
  • SafetyCap III, for Merck® bottles
    S40
    3× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €
  • SafetyCap I, for Merck® bottles with shut-off
    S40
    1× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 139,00 €
    139,00 €
    1 * 1 ST
       139,00 €
  • Safety Cap II for Wheaton bottles
    38/430
    2× 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  • SafetyCap I, V2.0, for Merck® bottles
    GL/S40
    1× PFA-fitting 1,6/2,3/3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 96,80 €
    96,80 €
    1 * 1 ST
       96,80 €
  • SafetyCap II, V2.0, for Merck® bottles
    GL/S40
    2× PFA-fitting 1,6/2,3/3,2 mm OD, 1× PFA-blind plug
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  • SafetyCap II, for ground neck bottles 29/32 mm, with locknut
    29/32 mm
    2× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
Angled
  • Description
    Thread
    Connector
  • SafetyCap I, V2.0, 90° angled
    GL45
    1× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • SafetyCap II, V2.0, 90° angled
    GL45
    2× PFA-fitting 3,2 mm OD
    1 * 1 ST
    1 * 1 ST - 139,00 €
    139,00 €
    1 * 1 ST
       139,00 €
Preperative
  •  
    Description
    Connector
    Thread
  • SafetyCap II, V2.0, preperative
    2× PTFE-fitting 6,35 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 172,00 €
    172,00 €
    1 * 1 ST
       172,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap II, V2.0, preperative
    1× PTFE-fitting 3,2 mm OD, 1× 6,35 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 151,00 €
    151,00 €
    1 * 1 ST
       151,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap I, V2.0, preperative
    1× PTFE-fitting 6,35 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap II, V2.0, preperative
    1× PFA-fitting 2,3/3,2 mm OD, 1× 4,76 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap I, V2.0, preperative
    1× PTFE-fitting 4,76 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap II, V2.0, preperative
    2× PTFE-fitting 4,76 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap III, V2.0, preperative
    3× PTFE-fitting 4,76 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
With shut-off
  •  
    Description
    Connector
    Thread
  • SafetyCap IV, V2.0, with 4× shut-off
    4× PFA-fittings 3,2 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap VI, V2.0, with 6× shut-off
    6× PFA-fitting 3,2 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 247,00 €
    247,00 €
    1 * 1 ST
       247,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap III, V2.0, with 3×shut-off
    3× PFA-fitting 3,2 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 150,00 €
    150,00 €
    1 * 1 ST
       150,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap II, V2.0, with 2× shut-off
    2× PFA-fitting 3,2 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 139,00 €
    139,00 €
    1 * 1 ST
       139,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap III, V2.0, with 2× shut-off (combined)
    3× PFA-fitting 3,2 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 150,00 €
    150,00 €
    1 * 1 ST
       150,00 €
  •  
    Description
    Connector
    Thread
  • SafetyCap II, V2.0, with shut-off (combined)
    2× PFA-fitting 3,2 mm OD
    GL45
    1 * 1 ST
    1 * 1 ST - 139,00 €
    139,00 €
    1 * 1 ST
       139,00 €
Replacement air valves
  •  
    Description
  • Air valve for SafetyCaps V 2.0, UNF ¹/₄" 28G, service life 6 months
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  •  
    Description
  • Air valve for SafetyCaps V 2.0, UNF ¹/₄" 28G, service life 6 months
    1 * 10 ST
    1 * 10 ST - 242,00 €
    242,00 €
    1 * 10 ST
       242,00 €
  •  
    Description
  • Air valve for SafetyCaps V 2.0, UNF ¹/₄" 28G, service life 6 months
    1 * 50 ST
    1 * 50 ST - 1 080,00 €
    1 080,00 €
    1 * 50 ST
       1 080,00 €
Replacement PFA fittings
  •  
    Description
    Material
    For tubing ext.Ø
    Colour
  • Fitting for capillary connector
    PFA
    2,3 mm
    Violet
    1 * 5 ST
    1 * 5 ST - 21,50 €
    21,50 €
    1 * 5 ST
       21,50 €
  •  
    Description
    Material
    For tubing ext.Ø
    Colour
  • Fitting for capillary connector
    PFA
    1,6 mm
    Green
    1 * 5 ST
    1 * 5 ST - 21,50 €
    21,50 €
    1 * 5 ST
       21,50 €
  •  
    Description
    Material
    For tubing ext.Ø
    Colour
  • Fitting for capillary connector
    PFA
    3,2 mm
    Blue
    1 * 5 ST
    1 * 5 ST - 21,50 €
    21,50 €
    1 * 5 ST
       21,50 €