Human recombinant prolactin R (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human Prolactin, Mouse IgG2a Fc Tag, low endotoxin at 1 μg/ml (100 µl/well) can bind Human Prolactin R, Fc Tag with a linear range of 2-39 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human GH, Tag Free at 2 μg/ml (100 µl/well) can bind Human Prolactin R, Fc Tag with a linear range of 0,4-6 ng/ml (Routinely tested).
- Gene ID:P16471-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:PRL-R|Prolactin R|PRLR
- Protein/peptide name:Prolactin R
- Purity:>95%
- Molecular weight:51,0 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Frequently Bought Together