Human recombinant BTN1A1 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Gene ID:Q13410
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:BTN|BTN1A1|Butyrophilin
- Protein/peptide name:BTN1A1
- Purity:>95%
- Molecular weight:50,6 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together