Rhesus macaque recombinant GITR (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Gene ID:F7DDV8
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD357|GITR|TNFRSF18|AITR
- Protein/peptide name:GITR
- Purity:>95%
- Molecular weight:41,3 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder, RT
Specifications
Frequently Bought Together