Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant transferrin R2 (from HEK293)
Mouse recombinant transferrin R2 (from HEK293)
Fournisseur:  ACROBIOSYSTEMS
Certificats
Mouse recombinant transferrin R2 (from HEK293)
Fournisseur:  ACROBIOSYSTEMS

Spécifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Gene ID:
    Q9JKX3-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    TFR2
  • Protein/peptide name:
    Transferrin R2
  • Purity:
    >95%
  • Molecular weight:
    101,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Spécifications

Frequently Bought Together