Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant SIRP alpha (from HEK293)
Mouse recombinant SIRP alpha (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse recombinant SIRP alpha (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBCD7-M525a) at 2 μg/ml (100 μl/well) can bind Mouse SIRP alpha, His Tag (Cat. No. ACRBSIA-M52H4) with a linear range of 0,02 - 0,625 μg/ml (QC tested)
  • Gene ID:
    P97797-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    MYD1|PTPNS1|CD172A|SHPS1|P84|SIRPA|BIT|MFR
  • Protein/peptide name:
    SIRP alpha
  • Purity:
    >95%
  • Molecular weight:
    39,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together