Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant PLGF (from HEK293)
Mouse recombinant PLGF (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Mouse recombinant PLGF (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 100 mM HAC, pH 2,5. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. Immobilised Mouse VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBVE1-M5256) at 2 μg/ml (100 μl/well) can bind Mouse PLGF, His Tag (Cat. No. ACRBPGF-M52H0) with a linear range of 0,2 - 4 ng/ml (QC tested).
  • Gene ID:
    P49764-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    PGF|PLGF|PlGF2|PGFL|PlGF|SHGC-10760
  • Protein/peptide name:
    PLGF
  • Purity:
    >95%
  • Molecular weight:
    17,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 100 mM HAC, pH 2,5. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together