Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant PD-L2 (from HEK293)
Mouse recombinant PD-L2 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Mouse recombinant PD-L2 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Biotinylated Human PD-1, Fc Tag (Cat. No. ACRBPD1-H82F1) at 1 μg/ml (100 μl/well) on streptavidin precoated (0,2 μg/well) plate, can bind Mouse PD-L2, His Tag (Cat. No. ACRBPD2-M52E3) with a linear range of 20 - 156 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. immobilised Mouse PD-L2, His Tag (Cat. No. ACRBPD2-M52E3) at 5 μg/ml (100 μl/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. ACRBPD1-M82F4) with a linear range of 0,156 - 2,5 μg/ml (Routinely tested).; Loaded Mouse PD-1, Fc Tag (Cat. No. ACRBPD1-M5259) on Protein A Biosensor, can bind Mouse PD-L2, His Tag (Cat. No. ACRBPD2-M52E3) with an affinity constant of 0,13 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested)
  • Gene ID:
    Q9WUL5
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    PDCD1L2|CD273|PD-L2|Butyrophilin B7-DC|PDL2|PDCD1LG2|PDCD1 ligand 2
  • Protein/peptide name:
    PD-L2
  • Purity:
    >95%
  • Molecular weight:
    24,4 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together