Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant BAFF (from HEK293)
Mouse recombinant BAFF (from HEK293)
Catalog # ACRBBAF-M521Y-50UG
Supplier:  ACRO Biosystems
CAS Number:  
Mouse recombinant BAFF (from HEK293)
Catalog # ACRBBAF-M521Y-50UG
Supplier:  ACRO Biosystems
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Seq:
    01
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Mouse BAFF, Tag Free (Cat. No. ACRBBAF-M521y) at 2 μg/ml (100 μl/well) can bind Mouse BAFFR, Fc Tag (Cat. No. ACRBBAR-M5259) with a linear range of 0,4 - 3 ng/ml (QC tested)
  • Gene ID:
    Q9WU72-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|BLYS|BAFF
  • Protein/peptide name:
    BAFF
  • Purity:
    >90%
  • Molecular weight:
    20,9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

About this item