Order Entry
Belgium
ContactUsLinkComponent
Mouse recombinant 4-1BB (from HEK293)
Mouse recombinant 4-1BB (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse recombinant 4-1BB (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation; Measured by its binding ability in a functional ELISA. immobilised Mouse 4-1BB, His Tag (Cat. No. ACRB41B-M52H7) at 0.5 μg/ml (100 μl/well) can bind Mouse 4‑1BB Ligand, Fc Tag (Cat. No. ACRB41L-M5257) with a linear range of 0,2 - 3 ng/ml (QC tested).; Loaded Mouse 4-1BB Ligand, Fc Tag (Cat. No. ACRB41L-M5257) on Protein A Biosensor, can bind Mouse 4-1BB, His Tag (Cat. No. ACRB41B-M52H7) with an affinity constant of 4,8 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested)
  • Gene ID:
    NP_001070976.1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CD137|TNFRSF9|ILA|CDw137|4-1BB
  • Protein/peptide name:
    4-1BB
  • Purity:
    >95%
  • Molecular weight:
    21,9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder, RT

Specifications

Frequently Bought Together