Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CD39 (from HEK293 cells), Biotin
Human recombinant CD39 (from HEK293 cells), Biotin
Catalog # ACRBCD9-H82E5-25UG
Supplier:  ACRO Biosystems
CAS Number:  
Human recombinant CD39 (from HEK293 cells), Biotin
Catalog # ACRBCD9-H82E5-25UG
Supplier:  ACRO Biosystems
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His, Avitag™
  • Storage conditions:
    –70 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    P49961-1
  • Protein synonyms:
    CD39|Ecto-ATPDase 1|Ecto-ATPase 1|ENTPD1|Entpd1|NTPDase 1
  • Protein/peptide name:
    CD39
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    54,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Delivered as bulk protein in a 0,2 μm filtered solution of 20 mM Tris, 150 mM NaCl, pH 8,0 with trehalose as protectant.
  • Shipping temperature:
    Dry Ice

Specifications

About this item

Biotinylated Human CD39 / ENTPD1 protein, His,Avitag™ (MALS verified) (active enzyme).