Human recombinant CD30 (from HEK293 cells), FITC (Fluorescein Isothiocyanate)
Supplier: ACRO Biosystems
Certificates
About this item
FITC-Labeled Human CD30 / TNFRSF8 protein, Fc Tag.
Specifications
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Type de protéine/peptide:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Conditions de stockage:–20 °C
- Teneur en endotoxines:Less than 1,0 EU per μg by the LAL method.
- Synonyme de protéine:Ki-1|CD30|D1S166E|TNFRSF8
- Nom de la protéine/du peptide:CD30
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:64,6 kDa
- Niveau d'endotoxine:Low
- Formulation:Please contact us for detailed information.
- Température de transport:RT
Specifications
Frequently Bought Together